Ethyl potassium malonate, 98%
Ethyl potassium malonate, 98%
Ethyl potassium malonate, 98%
Thermo Scientific Chemicals

Ethyl potassium malonate, 98%

CAS: 6148-64-7 | C5H7KO4 | 170.2 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 248080250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6148-64-7
IUPAC Name3-ethoxy-3-oxopropanoic acid potassium
Molecular FormulaC5H8KO4
InChI KeyHRBCMCJOBRIIQG-UHFFFAOYSA-N
SMILES[K].CCOC(=O)CC(O)=O
さらに表示
Appearance (Color)White to cream
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only