cis-2-Aminocyclohexanol hydrochloride, 95%
cis-2-Aminocyclohexanol hydrochloride, 95%
cis-2-Aminocyclohexanol hydrochloride, 95%
Thermo Scientific Chemicals

cis-2-Aminocyclohexanol hydrochloride, 95%

CAS: 6936-47-6 | C6H13NO·HCl | 151.64 g/mol
製品番号(カタログ番号) 265850010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6936-47-6
IUPAC Name(1S,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Molecular FormulaC6H13ClNO
InChI KeySEBSRZIHRZRNAH-IBTYICNHSA-N
SMILES[Cl].N[C@@H]1CCCC[C@@H]1O
さらに表示
Appearance (Form)Crystalline powder
Melting point186°C to 190°C
Infrared spectrumConforms
Appearance (Color)White to light beige
Titration with HClO494 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only