Benzyltrimethylammonium dichloroiodate, 99%
Benzyltrimethylammonium dichloroiodate, 99%
Benzyltrimethylammonium dichloroiodate, 99%
Thermo Scientific Chemicals

Benzyltrimethylammonium dichloroiodate, 99%

CAS: 114971-52-7 | C10H16Cl2IN | 348.04 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 268600250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS114971-52-7
IUPAC Namebenzyltrimethylazanium; dichloroiodanuide
Molecular FormulaC10H16Cl2IN
InChI KeyPPDJNZTUDFPAHX-UHFFFAOYSA-N
SMILESCl[I-]Cl.C[N+](C)(C)CC1=CC=CC=C1
さらに表示
Appearance (Form)Crystalline powder or flakes
Melting point125°C to 128°C
Appearance (Color)Yellow to light brown
Infrared spectrumConforms
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only