Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%
Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%

CAS: 6289-46-9 | C10H12O6 | 228.20 g/mol
数量:
50 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 271810500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6289-46-9
IUPAC Name1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Molecular FormulaC10H12O6
InChI KeyMHKKFFHWMKEBDW-UHFFFAOYNA-N
SMILESCOC(=O)C1CC(=O)C(CC1=O)C(=O)OC
さらに表示
Appearance (Color)White to yellow-green
HPLC>=98.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point153°C to 157°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only