2,3,5-Collidine, 99+%
2,3,5-Collidine, 99+%
2,3,5-Collidine, 99+%
Thermo Scientific Chemicals

2,3,5-Collidine, 99+%

CAS: 695-98-7 | C8H11N | 121.18 g/mol
製品番号(カタログ番号)数量パッケージ
27547025025 gGlass bottle
製品番号(カタログ番号) 275470250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS695-98-7
IUPAC Name2,3,5-trimethylpyridine
Molecular FormulaC8H11N
InChI KeyGFYHSKONPJXCDE-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C(C)=C1
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5065 to 1.5085 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only