alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
Thermo Scientific Chemicals

alpha-Bromo-2,6-difluorotoluene, 97%

CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 303570050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS85118-00-9
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point51°C to 55°C
Appearance (Form)Crystals and/or chunks
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only