1,8-Octanedithiol, 99%
1,8-Octanedithiol, 99%
1,8-Octanedithiol, 99%
Thermo Scientific Chemicals

1,8-Octanedithiol, 99%

CAS: 1191-62-4 | C8H18S2 | 178.35 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 310620010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1191-62-4
IUPAC Nameoctane-1,8-dithiol
Molecular FormulaC8H18S2
InChI KeyPGTWZHXOSWQKCY-UHFFFAOYSA-N
SMILESSCCCCCCCCS
さらに表示
Appearance (Form)Liquid
Refractive index1.5030 to 1.5050 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only