4-Tolylboronic acid, 97%
4-Tolylboronic acid, 97%
4-Tolylboronic acid, 97%
4-Tolylboronic acid, 97%
4-Tolylboronic acid, 97%
Thermo Scientific Chemicals

4-Tolylboronic acid, 97%

CAS: 5720-05-8 | C7H9BO2 | 135.96 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 344430050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5720-05-8
IUPAC Name(4-methylphenyl)boronic acid
Molecular FormulaC7H9BO2
InChI KeyBIWQNIMLAISTBV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)B(O)O
さらに表示
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Melting point256°C to 263°C
Appearance (Color)White to cream to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only