3-Bromophenethyl alcohol, 97%
3-Bromophenethyl alcohol, 97%
3-Bromophenethyl alcohol, 97%
Thermo Scientific Chemicals

3-Bromophenethyl alcohol, 97%

CAS: 28229-69-8 | C8H9BrO | 201.06 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 350560050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS28229-69-8
Refractive index1.5730 to 1.5750 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only