4-Hydroxy-3,5-dimethylbenzoic acid, 98%
4-Hydroxy-3,5-dimethylbenzoic acid, 98%
4-Hydroxy-3,5-dimethylbenzoic acid, 98%
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzoic acid, 98%

CAS: 4919-37-3 | C9H10O3 | 166.18 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 351760250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4919-37-3
IUPAC Name4-hydroxy-3,5-dimethylbenzoate
Molecular FormulaC9H9O3
InChI KeyOMNHTTWQSSUZHO-UHFFFAOYSA-M
SMILESCC1=CC(=CC(C)=C1O)C([O-])=O
さらに表示
Appearance (Color)Off-white to light beige to light pink
Infrared spectrumConforms
Melting point221°C to 226°C
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only