Glycerine trioleate, 98%
Glycerine trioleate, 98%
Glycerine trioleate, 98%
Thermo Scientific Chemicals

Glycerine trioleate, 98%

CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
数量:
1 mL
5 mL
25 mL
100 mL
製品番号(カタログ番号) 368120250
価格(JPY)
-
数量:
25 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS122-32-7
IUPAC Name1,3-bis[(9Z)-octadec-9-enoyloxy]propan-2-yl (9Z)-octadec-9-enoate
Molecular FormulaC57H104O6
InChI KeyPHYFQTYBJUILEZ-IUPFWZBJSA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
さらに表示
Refractive index1.4680 to 1.4700 (20°C, 589 nm)
Appearance (Form)Liquid
HPLC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only