Pinacolborane, 97%, stabilized
Pinacolborane, 97%, stabilized
Pinacolborane, 97%, stabilized
Thermo Scientific Chemicals

Pinacolborane, 97%, stabilized

CAS: 25015-63-8 | C6H13BO2 | 127.98 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 371631000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25015-63-8
IUPAC Name4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC6H13BO2
InChI KeyUCFSYHMCKWNKAH-UHFFFAOYSA-N
SMILESCC1(C)OBOC1(C)C
さらに表示
Stabilizer0.1 to 1.5 % Triethylamine
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3950 to 1.4000 (20°C, 589 nm)
GC>=96.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only