N-Phenylbis(trifluoromethanesulfonimide), 97%
N-Phenylbis(trifluoromethanesulfonimide), 97%
N-Phenylbis(trifluoromethanesulfonimide), 97%
Thermo Scientific Chemicals

N-Phenylbis(trifluoromethanesulfonimide), 97%

CAS: 37595-74-7 | C8H5F6NO4S2 | 357.25 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 374790250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS37595-74-7
IUPAC Name1,1,1-trifluoro-N-phenyl-N-trifluoromethanesulfonylmethanesulfonamide
Molecular FormulaC8H5F6NO4S2
InChI KeyDIOHEXPTUTVCNX-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)N(C1=CC=CC=C1)S(=O)(=O)C(F)(F)F
さらに表示
Melting point99°C to 105°C
HPLC>=96.0 %
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only