2-Octyn-1-ol, 98%
2-Octyn-1-ol, 98%
2-Octyn-1-ol, 98%
Thermo Scientific Chemicals

2-Octyn-1-ol, 98%

CAS: 20739-58-6 | C8H14O | 126.20 g/mol
数量:
5 mL
25 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 387030050
価格(JPY)
-
数量:
5 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20739-58-6
IUPAC Nameoct-2-yn-1-ol
Molecular FormulaC8H14O
InChI KeyTTWYFVOMGMBZCF-UHFFFAOYSA-N
SMILESCCCCCC#CCO
さらに表示
Appearance (Form)Liquid
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only