4'-Iodoacetanilide, 97%
4'-Iodoacetanilide, 97%
4'-Iodoacetanilide, 97%
Thermo Scientific Chemicals

4'-Iodoacetanilide, 97%

CAS: 622-50-4 | C8H8INO | 261.06 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 393120010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS622-50-4
IUPAC NameN-(4-iodophenyl)acetamide
Molecular FormulaC8H8INO
InChI KeySIULLDWIXYYVCU-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(I)C=C1
さらに表示
Appearance (Color)Bleu-grey to purple
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only