3-Bromo-2-butanone, 95%, stabilized
3-Bromo-2-butanone, 95%, stabilized
3-Bromo-2-butanone, 95%, stabilized
Thermo Scientific Chemicals

3-Bromo-2-butanone, 95%, stabilized

CAS: 814-75-5 | C4H7BrO | 151 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 402910010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS814-75-5
IUPAC Name3-bromobutan-2-one
Molecular FormulaC4H7BrO
InChI KeyBNBOUFHCTIFWHN-UHFFFAOYNA-N
SMILESCC(Br)C(C)=O
さらに表示
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
Stabilizer=<1 % magnesium oxide
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only