Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals
Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals
Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals
Thermo Scientific Chemicals

Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals

CAS: 7790-84-3 | Cd3O12S3·8H2O | 769.51 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 403761000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7790-84-3
IUPAC Namecadmium(2+) hydrate sulfate
Molecular FormulaCdH2O5S
InChI KeyVRFRVKRTIMIRNG-UHFFFAOYSA-L
SMILESO.[Cd++].[O-]S([O-])(=O)=O
さらに表示
Appearance (Form)Crystals
Nitrate and Nitrite (as NO3)=<0.003 %
Chloride (Cl)=<0.001 %
Copper (Cu)=<0.002 %
Iron (Fe)=<0.001 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only