chlorodifluoroacetic acid sodium salt, 96%
chlorodifluoroacetic acid sodium salt, 96%
chlorodifluoroacetic acid sodium salt, 96%
Thermo Scientific Chemicals

chlorodifluoroacetic acid sodium salt, 96%

CAS: 1895-39-2 | C2ClF2NaO2 | 152.46 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 404590050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1895-39-2
IUPAC Namesodium 2-chloro-2,2-difluoroacetate
Molecular FormulaC2ClF2NaO2
InChI KeyMRTAVLDNYYEJHK-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C(F)(F)Cl
さらに表示
Infrared spectrumConforms
Titration with HClO4>=95 %
Appearance (Form)Powder
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only