Guanosine, 99%
Guanosine, 99%
Guanosine, 99%
Thermo Scientific Chemicals

Guanosine, 99%

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 411135000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS118-00-3
IUPAC Name2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H13N5O5
InChI KeyNYHBQMYGNKIUIF-UUOKFMHZSA-N
SMILESNC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
さらに表示
Titration with HClO4>=98.5 to 101.5 % (On dry substance)
Appearance (Form)Powder
Appearance (Color)White to light yellow
Loss on drying=<5 % (1 g, 105°C, 3 h)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only