Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Thermo Scientific Chemicals

Hexadecyltrimethylammonium hydroxide, 25% in methanol

CAS: 505-86-2 | C19H43NO | 301.56 g/mol
数量:
50 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 411420500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS505-86-2
IUPAC Namehexadecyltrimethylazanium hydroxide
Molecular FormulaC19H43NO
InChI KeyWJLUBOLDZCQZEV-UHFFFAOYSA-M
SMILES[OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C
さらに表示
Water=<3 % (K.F.)
Concentration23 to 27 w/w% (in methanol)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only