Tetrapropylammonium Hydroxide, 10% in Water
Tetrapropylammonium Hydroxide, 10% in Water
Tetrapropylammonium Hydroxide, 10% in Water
Thermo Scientific Chemicals

Tetrapropylammonium Hydroxide, 10% in Water

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 420721000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4499-86-9
IUPAC Nametetrapropylazanium hydroxide
Molecular FormulaC12H29NO
InChI KeyLPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES[OH-].CCC[N+](CCC)(CCC)CCC
さらに表示
Appearance (Color)Clear colorless
Titration with HClO49 to 11 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only