Trimethylamine N-oxide dihydrate, 98%
Trimethylamine N-oxide dihydrate, 98%
Trimethylamine N-oxide dihydrate, 98%
Thermo Scientific Chemicals

Trimethylamine N-oxide dihydrate, 98%

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 421895000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62637-93-8
IUPAC NameN,N-dimethylmethanamine oxide dihydrate
Molecular FormulaC3H13NO3
InChI KeyPGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILESO.O.C[N+](C)(C)[O-]
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point94°C to 98°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only