Hexaethylphosphorus Triamide, 95%
Hexaethylphosphorus Triamide, 95%
Hexaethylphosphorus Triamide, 95%
Hexaethylphosphorus Triamide, 95%
Thermo Scientific Chemicals

Hexaethylphosphorus Triamide, 95%

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 423080050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2283-11-6
IUPAC Name[bis(diethylamino)phosphanyl]diethylamine
Molecular FormulaC12H30N3P
InChI KeyFDIOSTIIZGWENY-UHFFFAOYSA-N
SMILESCCN(CC)P(N(CC)CC)N(CC)CC
さらに表示
Refractive index1.4720 to 1.4750 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only