Reinecke Salt, ACS Reagent
Reinecke Salt, ACS Reagent
Reinecke Salt, ACS Reagent
Thermo Scientific Chemicals

Reinecke Salt, ACS Reagent

CAS: 13573-16-5 | C4H10CrN7S4·H2O | 354.46 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 423335000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13573-16-5
IUPAC Namechromium(3+) ammonium tetrakis(cyanosulfanide) diamine
Molecular FormulaC4H10CrN7S4
InChI KeyZGLIQORZYPZFPW-UHFFFAOYSA-K
SMILESN.N.[NH4+].[Cr+3].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
さらに表示
Appearance (Color)Red
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Insoluble matter=<0.05 % (in dilute HCl)
Sensitivitypasses test
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only