Methyl tricosanoate, 99+%
Methyl tricosanoate, 99+%
Methyl tricosanoate, 99+%
Thermo Scientific Chemicals

Methyl tricosanoate, 99+%

CAS: 2433-97-8 | C24H48O2 | 368.65 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 428450010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2433-97-8
Infrared spectrumConforms
Melting point53°C to 56°C
Appearance (Color)White
Appearance (Form)Powder or solid
GC>=99.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only