3-(BOC-Amino)piperidine, 97%
3-(BOC-Amino)piperidine, 97%
3-(BOC-Amino)piperidine, 97%
Thermo Scientific Chemicals

3-(BOC-Amino)piperidine, 97%

CAS: 172603-05-3 | C10H20N2O2 | 200.28 g/mol
数量:
1 g
5 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 437510010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS172603-05-3
IUPAC Nametert-butyl N-(piperidin-3-yl)carbamate
Molecular FormulaC10H20N2O2
InChI KeyWUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC1CCCNC1
さらに表示
Appearance (Form)Crystalline powder
GC>=96.0 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only