4-Aminocarbonylphenylboronic acid, 97%
4-Aminocarbonylphenylboronic acid, 97%
4-Aminocarbonylphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Aminocarbonylphenylboronic acid, 97%

CAS: 123088-59-5 | C7H8BNO3 | 164.96 g/mol
製品番号(カタログ番号) 444900050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123088-59-5
IUPAC Name(4-carbamoylphenyl)boronic acid
Molecular FormulaC7H8BNO3
InChI KeyGNRHNKBJNUVWFZ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C(C=C1)B(O)O
さらに表示
HPLC>=96.0 %
Appearance (Color)White to tan
Appearance (Form)Solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only