1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%

CAS: 864754-16-5 | C13H21BN2O4 | 280.13 g/mol
製品番号(カタログ番号) 445010010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS864754-16-5
IUPAC Nameethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetate
Molecular FormulaC13H21BN2O4
InChI KeyYUEZJHOSHBTWPV-UHFFFAOYSA-N
SMILESCCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
さらに表示
Appearance (Color)Colorless to yellow to orange
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only