Sodium nitrate, 99+%, for biochemistry
Sodium nitrate, 99+%, for biochemistry
Sodium nitrate, 99+%, for biochemistry
Thermo Scientific Chemicals

Sodium nitrate, 99+%, for biochemistry

CAS: 7631-99-4 | NNaO3 | 84.99 g/mol
数量:
250 g
1 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 446810010
価格(JPY)
-
数量:
1 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7631-99-4
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
さらに表示
pH5.5 to 8.3
Sulfate (SO4)=<0.003 %
Insoluble matter=<0.005 %
Appearance (Color)Colorless to white
Titration after Ion exchange>=99.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only