Citric acid, diammonium salt, 98+%, ACS reagent
Citric acid, diammonium salt, 98+%, ACS reagent
Citric acid, diammonium salt, 98+%, ACS reagent
Thermo Scientific Chemicals

Citric acid, diammonium salt, 98+%, ACS reagent

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol
数量:
100 g
500 g
2.5 kg
製品番号(カタログ番号) 447680025
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
さらに表示
Titration with NaOH98.0 to 103.0 %
Appearance (Form)Crystalline powder
Heavy metals (ICP-OES)=<5 ppm
Iron (Fe)=<0.001 %
Oxalate (C2O4)Passes test (Limit about 0.05%)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only