2'-Deoxycytidine hydrochloride, 98%
2'-Deoxycytidine hydrochloride, 98%
2'-Deoxycytidine hydrochloride, 98%
Thermo Scientific Chemicals

2'-Deoxycytidine hydrochloride, 98%

CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol
製品番号(カタログ番号) 458510010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3992-42-5
IUPAC Name4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
Molecular FormulaC9H14ClN3O4
InChI KeyLTKCXZGFJFAPLY-VOBHXVAMNA-N
SMILESCl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
さらに表示
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
HPLC>=97.5 %
Water=<8 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only