Pyridostigmine bromide, 98%
Pyridostigmine bromide, 98%
Pyridostigmine bromide, 98%
Thermo Scientific Chemicals

Pyridostigmine bromide, 98%

CAS: 101-26-8 | C9H13BrN2O2 | 261.12 g/mol
製品番号(カタログ番号) 461190010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS101-26-8
IUPAC Name3-[(dimethylcarbamoyl)oxy]-1-methylpyridin-1-ium bromide
Molecular FormulaC9H13BrN2O2
InChI KeyVNYBTNPBYXSMOO-UHFFFAOYSA-M
SMILES[Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1
さらに表示
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point153°C to 157°C
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only