Potassium hydrogen phthalate, for HPLC
Thermo Scientific Chemicals

Potassium hydrogen phthalate, for HPLC

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol
製品番号(カタログ番号) 463850500
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
さらに表示
Infrared spectrumConforms
pH3.5 to 5.0 (5 % in water)
Solubility(10 % in water) Clear colorless
UVat 330 nm A: =<0.05
Appearance (Form)Crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only