1,4-Benzenedimethanethiol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Benzenedimethanethiol, Thermo Scientific Chemicals

製品番号(カタログ番号) 467610010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS105-09-9
IUPAC Name[4-(sulfanylmethyl)phenyl]methanethiol
Molecular FormulaC8H10S2
InChI KeyIYPNRTQAOXLCQW-UHFFFAOYSA-N
SMILESSCC1=CC=C(CS)C=C1
さらに表示
Melting point40°C to 45°C
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder, crystals or solid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only