2,3,4-Trifluoroaniline, 98%
2,3,4-Trifluoroaniline, 98%
2,3,4-Trifluoroaniline, 98%
Thermo Scientific Chemicals

2,3,4-Trifluoroaniline, 98%

CAS: 3862-73-5 | C6H4F3N | 147.1 g/mol
製品番号(カタログ番号) A10254.14
または、製品番号A10254-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3862-73-5
IUPAC Name2,3,4-trifluoroaniline
Molecular FormulaC6H4F3N
InChI KeyWRDGNXCXTDDYBZ-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C(F)=C1F
さらに表示
FormLiquid
Appearance (Color)Clear colorless to yellow to dark red to pale brown
Refractive Index1.4845-1.4885 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only