Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%
Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%
Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%
Thermo Scientific Chemicals

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A10276.30
または、製品番号A10276-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS334024-15-6
IUPAC Nameiron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC6H5FeO7
InChI KeyNPFOYSMITVOQOS-UHFFFAOYSA-K
SMILES[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
さらに表示
Appearance (Color)Orange to brown
FormPowder
Assay from Suppliers CofAFe(II) content ≤5.0%
Assay from Suppliers CofAFe(III) content ≥16.5% to ≤20.0%
Water Content (Karl Fischer Titration)≤27.0%
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Itzhak Shweky; Avi Bino; David P. Goldberg; Stephen J. Lippard. Syntheses, Structures, and Magnetic Properties of Two Dinuclear Iron(III) Citrate Complexes. Inorg. Chem. 1994, 33, (23), 5161-5162.