Thermo Scientific Chemicals

2-(Methylthio)benzoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A10379.14
25 g, Each
Thermo Scientific Chemicals

2-(Methylthio)benzoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A10379.14
25 g, Each
数量
製品番号(カタログ番号): A10379.14
または、製品番号A10379-14

化学物質識別子

CAS
3724-10-5
IUPAC Name
2-(methylsulfanyl)benzoic acid
Molecular Formula
C8H8O2S
InChI Key
LWJQGKJCZOGGPJ-UHFFFAOYSA-N
SMILES
CSC1=CC=CC=C1C(O)=O
Assay (Silylated GC)
≥98.0%
Melting Point (clear melt)
166-171?C
Appearance (Color)
White to cream
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

概要

2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Soluble in Methanol.

Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text