Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water
Thermo Scientific Chemicals

Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10510.22
または、製品番号A10510-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7787-60-2
IUPAC Namebismuth(3+) trichloride
Molecular FormulaBiCl3
InChI KeyJHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Bi+3]
さらに表示
Assay (Titration ex Chloride)≥97.0 to 103.0% (dry weight)
Loss on Drying≤3.0%
Appearance (Color)White to cream
Elemental AnalysisBismuth Bi (dry weight): 64.28-68.26% (Theory 66.27%)
FormPowder
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

Solubility
Soluble in hydrochloric acid, nitric acid, diethyl ether, ethyl acetate and acetone. Insoluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Lewis acid catalyst which promotes the rearrangement of acetanilides to o-amino ketones: Tetrahedron Lett., 3217 (1976).
  2. Silyl enol ethers can be ɑ-acylated with acyl chlorides in the presence of a catalytic amount of BiCl3 and NaI or ZnI2; other Lewis acids such as ZnCl2 were less effective: J. Org. Chem., 61, 3885 (1996).
  3. Catalyzes the addition of 1,3-dicarbonyl compounds to Michael acceptors under solvent-free conditions with microwave irradiation: Tetrahedron Lett., 38, 1449 (1997).
  4. For catalysis of Diels-Alder and hetero Diels-Alder reactions, see: J. Org. Chem., 62, 4880 (1997); Tetrahedron Lett., 39, 1161 (1998).
  5. Arenes undergo sufonylation by heating with arenesulfonyl chlorides in the presence of BiCl3 (5 mol%) and triflic acid (10 mol%) to give high yields of mixed diaryl sulfones; neither BiCl3 nor TfOH alone is effective: Tetrahedron Lett., 40, 9233 (1999).
  6. Promotes the chemoselective deprotection of N-Boc amino acids and peptides: Tetrahedron Lett., 47, 389 (2006).
  7. For a brief feature on Bi(III) catalysts, see: Synlett, 1194 (2001). For a review of their use in acylation and sulfonylation reactions, see: Synlett, 181 (2002).
  8. Antonio, M. R.; McAlister, D. R.; Horwitz, E. P. An europium(III) diglycolamide complex: insights into the coordination chemistry of lanthanides in solvent extraction. Dalton Trans. 2015, 44 (2), 515-521.
  9. Vuorinen, S.; Lahcini, M.; Hatanpää, T.; Sundberg, M.; Leskelä, M.; Repo, T. Bismuth(III) Alkoxide Catalysts for Ring-Opening Polymerization of Lactides and ε-Caprolactone. Macromol. Chem. Phys. 2013, 214 (6), 707-715.