2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
Thermo Scientific Chemicals

2,4-Dichloroaniline, 99%

CAS: 554-00-7 | C6H5Cl2N | 162.013 g/mol
数量:
100 g
250 g
500 g
2500 g
製品番号(カタログ番号) A10518.30
または、製品番号A10518-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS554-00-7
IUPAC Name2,4-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyKQCMTOWTPBNWDB-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1Cl
さらに表示
Identification (FTIR)Conforms
Appearance (Color)White to cream to grey or pale brown to brown
FormPowder or crystalline powder or crystals or flakes and/or chunks
Melting Point (clear melt)58-67°C
CommentMay darken on storage
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only