Thermo Scientific Chemicals

Methyl 4-(bromomethyl)benzoate, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A10557.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-(bromomethyl)benzoate, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A10557.14
25 g, Each
数量
製品番号(カタログ番号): A10557.14
または、製品番号A10557-14

化学物質識別子

CAS
2417-72-3
Assay (GC)
≥97.5%
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
52.0-59.0°C

概要

Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

Solubility
Lowly soluble in water, highly soluble in ethanol and ether, not soluble in halogenated organic solvents

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases. Stable under recommended storage conditions.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text