Potassium hexafluorotitanate, 97%
Potassium hexafluorotitanate, 97%
Potassium hexafluorotitanate, 97%
Thermo Scientific Chemicals

Potassium hexafluorotitanate, 97%

CAS: 16919-27-0 | F6K2Ti
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A10915.22
または、製品番号A10915-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16919-27-0
IUPAC Nametitanium(4+) dipotassium hexafluoride
Molecular FormulaF6K2Ti
InChI KeyRXCBCUJUGULOGC-UHFFFAOYSA-H
SMILES[F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Ti+4]
さらに表示
Appearance (Color)White
Elemental AnalysisPotassium K : 31.27 - 33.87% (Theory 32.56%)
Elemental AnalysisTitanium Ti: 19.14 - 20.73% (Theory 19.94%)
Formpowder
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mansoor, M.; Shahid, M. A facile one-step method of coating aluminum on multiwall carbon nanotubes. J. Alloys Compd. 2015, 643, 74-78.
  2. Chen, C. G.; Luo, J.; Guo, Z. M.; Yang, W. W.; Chen, J. Microstructural evolution and mechanical properties of in situ TiB2/Al composites under high-intensity ultrasound. Rare Met. 2015, 34 (3), 168-172.