2,4,6-Trifluoroaniline, 98%
2,4,6-Trifluoroaniline, 98%
2,4,6-Trifluoroaniline, 98%
Thermo Scientific Chemicals

2,4,6-Trifluoroaniline, 98%

CAS: 363-81-5 | C6H4F3N | 147.1 g/mol
数量:
1 g
5 g
25 g
100 g
製品番号(カタログ番号) A10964.22
または、製品番号A10964-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS363-81-5
IUPAC Name2,4,6-trifluoroaniline
Molecular FormulaC6H4F3N
InChI KeyBJSVKBGQDHUBHZ-UHFFFAOYSA-N
SMILESNC1=C(F)C=C(F)C=C1F
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)31.0-37.0?C
Assay (GC)≥97.5%
Appearance (Color)White to pale yellow
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N?-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N′-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Karl Haider; N.Soundararajan; Myron Shaffer; Matthew S.Platz. EPR spectroscopy of a diaza derivative of meta-xylylene. Tetrahedron Letters. 1989, 30,(10), 1225-1228.
  2. P.Vallat; W.Fan; N.El Tayar; P.A.Carrupt; B.Testa. Solvatochromic analysis of the retention mechanism of two novel stationary phases used for measuring lipophilicity by RP-HPLC. Journal of Liquid Chromatography. 1992, 15,(12), 2133-2151.