Fumaric acid, 99%
Fumaric acid, 99%
Fumaric acid, 99%
Thermo Scientific Chemicals

Fumaric acid, 99%

CAS: 110-17-8 | C4H4O4 | 116.07 g/mol
数量:
10,000 g
500 g
2500 g
製品番号(カタログ番号) A10976.36
または、製品番号A10976-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-17-8
IUPAC Name(2E)-but-2-enedioic acid
Molecular FormulaC4H4O4
InChI KeyVZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILESOC(=O)\C=C\C(O)=O
さらに表示
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay (Silylated GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms
さらに表示
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.

Notes
Incompatible with oxidizing agents, amines and strong bases.
RUO – Research Use Only

General References:

  1. Rose, J. L.; Tata, B. V. R.; Talmon, Y.; Aswal, V. K.; Hassan, P. A.; Sreejith, L. Distinctive effect of maleic acid and fumaric acid on structural transitions in cationic micellar solution. Colloid. Polym. Sci. 2015, 293 (5), 1383-1390.
  2. Huang, D.; Wang, R.; Du, W.; Wang, G.; Xia, M. Activation of glycerol metabolic pathway by evolutionary engineering of Rhizopus oryzae to strengthen the fumaric acid biosynthesis from crude glycerol. Bioresour. Technol. 2015, 196, 263-272.