3,5-Dichlorobenzenesulfonyl chloride, 97%
3,5-Dichlorobenzenesulfonyl chloride, 97%
3,5-Dichlorobenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

3,5-Dichlorobenzenesulfonyl chloride, 97%

CAS: 705-21-5 | C6H3Cl3O2S | 245.50 g/mol
製品番号(カタログ番号) A11003.06
または、製品番号A11003-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS705-21-5
IUPAC Name3,5-dichlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3Cl3O2S
InChI KeyRJSQINMKOSOUGT-UHFFFAOYSA-N
SMILESClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O
さらに表示
Melting Point (clear melt)28.0-37.0?C
Appearance (Color)White to pale brown or clear colorless to pale yellow
FormFused solid or crystals or liquid as melt
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only