Benzonitrile, 99%
Benzonitrile, 99%
Benzonitrile, 99%
Thermo Scientific Chemicals

Benzonitrile, 99%

CAS: 100-47-0 | C7H5N | 103.124 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A11173.22
または、製品番号A11173-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-47-0
IUPAC Namebenzonitrile
Molecular FormulaC7H5N
InChI KeyJFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1
さらに表示
Assay from Supplier's CofA≥98.5% (US-Sourced Material)
Refractive Index1.5275-1.5305 @ 20?C (UK-Sourced Material)
Appearance (Color)Clear colorless
Assay (GC)≥98.5% (UK-Sourced Material)
CommentMaterial Sourced in UK and US
さらに表示
Benzonitrile is a widely utilized as a solvent and an intermediate in industries making drugs, perfumes, dyes, rubber, textiles, resins and specialty lacquers. It finds application as a versatile precursor for many derivatives. It coordinates with transition metal to form complexes which act as synthetic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzonitrile is a widely utilized as a solvent and an intermediate in industries making drugs, perfumes, dyes, rubber, textiles, resins and specialty lacquers. It finds application as a versatile precursor for many derivatives. It coordinates with transition metal to form complexes which act as synthetic intermediates.

Solubility
Miscible with common organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents, reducing agents, acids, chlorates, nitrates and plastics.
RUO – Research Use Only

General References:

  1. Patterson, D.; Doyle, J. M. A slow, continuous beam of cold benzonitrile. Phys. Chem. Chem. Phys. 2015, 17 (7), 5372-5375.
  2. Li, L.; Zou, J. Y.; You, S. Y.; Hu, Y.; Ji, Q. R.; Chen, K. H.; Cui, J. Z. A fluorescence chemosensor for benzonitrile derived from in-situ synthesis of azolate-carboxylic acid and zinc(II) ions. Inorg. Chem. Commun. 2015, 60, 111-114.