Tetra-n-propylammonium bromide, 98%
Tetra-n-propylammonium bromide, 98%
Tetra-n-propylammonium bromide, 98%
Thermo Scientific Chemicals

Tetra-n-propylammonium bromide, 98%

CAS: 1941-30-6 | C12H28BrN | 266.267 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A11522.36
または、製品番号A11522-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1941-30-6
IUPAC Nametetrapropylazanium bromide
Molecular FormulaC12H28BrN
InChI KeyBGQMOFGZRJUORO-UHFFFAOYSA-M
SMILES[Br-].CCC[N+](CCC)(CCC)CCC
さらに表示
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or lumps
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
RUO – Research Use Only

General References:

  1. Kyrre Thalberg.; Bjoern Lindman. Interaction between hyaluronan and cationic surfactants. J. Phys. Chem. 1989, 93, (4), 1478-1483.
  2. Herbert O. House.; Edith Feng.; Norton P. Peet. Comparison of various tetraalkylammonium salts as supporting electrolytes in organic electrochemical reactions. J. Org. Chem. 1971, 36, (16), 2371-2375.