Boron oxide, 98%
Boron oxide, 98%
Boron oxide, 98%
Thermo Scientific Chemicals

Boron oxide, 98%

CAS: 1303-86-2 | B2O3 | 69.62 g/mol
製品番号(カタログ番号)数量
A11707.0E
または、製品番号A11707-0E
2500 g
製品番号(カタログ番号) A11707.0E
または、製品番号A11707-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1303-86-2
IUPAC Nameoxo[(oxoboranyl)oxy]borane
Molecular FormulaB2O3
InChI KeyJKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILESO=BOB=O
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥97.5%
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kline, J.; Tangstad, M.; Tranell, G. A Raman Spectroscopic Study of the Structural Modifications Associated with the Addition of Calcium Oxide and Boron Oxide to Silica. Metall. Mater. Trans. B 2015, 46 (1), 62-73.
  2. Solozhenko, V. L.; Kurakevych, O. O.; Godec, Y. L.; Brazhkin, V. V. Thermodynamically Consistent p-T Phase Diagram of Boron Oxide B2O3 by in Situ Probing and Thermodynamic Analysis. J. Phys. Chem. C 2015, 119 (35), 20600-20605.