6-Methoxyquinoline, 98%
6-Methoxyquinoline, 98%
6-Methoxyquinoline, 98%
Thermo Scientific Chemicals

6-Methoxyquinoline, 98%

CAS: 5263-87-6 | C10H9NO | 159.188 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A11736.06
または、製品番号A11736-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5263-87-6
IUPAC Name6-methoxyquinoline
Molecular FormulaC10H9NO
InChI KeyHFDLDPJYCIEXJP-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=CC=CC2=C1
さらに表示
Assay (GC)≥97.5%
FormLiquid or low melting solid
Appearance (Color)Clear colorless to yellow to orange or pink to purple to brown
Refractive Index1.6200-1.6260 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only