Methyl 3,4,5-trimethoxybenzoate, 98+%
Methyl 3,4,5-trimethoxybenzoate, 98+%
Methyl 3,4,5-trimethoxybenzoate, 98+%
Thermo Scientific Chemicals

Methyl 3,4,5-trimethoxybenzoate, 98+%

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A11933.14
または、製品番号A11933-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1916-07-0
IUPAC Namemethyl 3,4,5-trimethoxybenzoate
Molecular FormulaC11H14O5
InChI KeyKACHFMOHOPLTNX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1
さらに表示
Appearance (Color)White to cream
Melting Point (clear melt)79.0-87.0°C
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only