Dimethoxydimethylsilane, 97%
Dimethoxydimethylsilane, 97%
Dimethoxydimethylsilane, 97%
Thermo Scientific Chemicals

Dimethoxydimethylsilane, 97%

CAS: 1112-39-6 | C4H12O2Si | 120.22 g/mol
製品番号(カタログ番号) A12025.22
または、製品番号A12025-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1112-39-6
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.3685-1.3735 @ 20?C
It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

Solubility
Contact with water releases a flammable alcohol.

Notes
Moisture Sensitive. Store away from oxidizing agents and water/moisture. Protect against electrostatic charges. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Nozomu Takano.; Tomio Fukuda and Katsumichi Ono. Analysis of Structures of Oligomeric Siloxanes from Dimethoxydimethylsilane under Heat Treatment by FT-IR.Polymer Journal.2001,33469-474.
  2. Günther Maier and Jörg Glatthaar. Analysis of Dimethoxydimethylsilane from Silicon Atoms and Dimethyl Ether: A Combined Matrix-Spectroscopic and Density Functional Theory Study.Eur. J. Org. Chem.2003,2003(17), 3350-3363.
  3. Silylating agent for diols.